L (+)-Vinsyra, EMPROVE® ESSENTIAL BP, E334, Ph. Eur., JP, NF, extra pure suitable for use as excipient, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Fara
Synonyms: L-Tartaric Acid, (2R,3R)-(+)-Tartaric acid, Tartaric Acid, (R,R)-(+)-2,3-Dihydroxysuccinic acid, (R,R)-(+)-Tartaric acid, (2R,3R)-2,3-Dihydroxysuccinic acid
1.00802.5000 1.00802.9029
1.00802.5000EA 21850 SEK
1.00802.5000 1.00802.9029
L (+)-Vinsyra, EMPROVE® ESSENTIAL BP, E334, Ph. Eur., JP, NF, extra pure suitable for use as excipient, SAFC®
L (+)-Vinsyra
Formel: C₄H₆O₆
Kokpunkt: 399 °C (1013 hPa)
Smältpunkt: 168…170 °C
Densitet: 1,759 g/cm³ (20 °C)
Flampunkt: 150 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00064207
CAS nummer: 87-69-4
EINECS: 201-766-0
REACH: 01-2119537204-47
Merck Index: 13,09158
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4e859e52081482a351d52322717614aad450742&vid=879c292903ddee7a59d5d6b2d969c4c8fc6eac75&prid=web&clid=SAPDAM