L (+)-Vinsyra ≥99%
Leverantör: Thermo Fisher Scientific
Fara
A13668.22 A13668.0E A13668.36
A13668.22EA 575 SEK
A13668.22 A13668.0E A13668.36
L (+)-Vinsyra ≥99%
L (+)-Vinsyra
Purity: 99% CAS No.: 87-69-4 Molecular Formula: C4H6O6 Formula Weight: 150.09 Odour: Odourless pH: 2.1 Melting Point: 168-170° Beilstein Reference: 1725147 Merck Reference: 14,9070 Optical Rotation: +12.4° (c=20 in water) MDL No.: MFCD00064207
Formel: C₄H₆O₆
Kokpunkt: 399 °C (1013 hPa)
Smältpunkt: 168…170 °C
Densitet: 1,760
Flampunkt: 150 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00064207
CAS nummer: 87-69-4
EINECS: 201-766-0
REACH: 01-2119537204-47
Merck Index: 14,9070
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4e859e52081482a351d52322717614aad450742&vid=879c292903ddee7a59d5d6b2d969c4c8fc6eac75&prid=web&clid=SAPDAM