L (+)-Asparaginsyra ≥99% low metals
Leverantör: Thermo Scientific
Synonyms: (S)-(+)-Aspartic acid, L(+)-2-Amino-1,4-butanedioic acid, (S)-(+)-2-Amino-1,4-butanedioic acid, H-Asp-OH, L-α-Aminosuccinic acid, L-αlpha-Aminosuccinic acid, (S)-2-Aminosuccinic acid, Asp, D
43317.22 43317.36
43317.22EA 710 SEK
43317.22 43317.36
L (+)-Asparaginsyra ≥99% low metals
L(+)-Aspartic acid
Purity: 99% CAS No.: 56-84-8 Molecular Formula: C4H7NO4 Formula Weight: 133.10 Physical Form: Powder Density: 1.66 Melting Point: 300° dec. Beilstein Reference: 1723530 Merck Reference: 14,840 MDL No.: MFCD00002616
Formel: C₄H₇NO₄
MW: 133,1 g/mol
Kokpunkt: 311 °C (1013 hPa)
Smältpunkt: >300 °C (dec.)
Densitet: 1,66
Storage Temperature: Omgivande
MDL Number: MFCD00002616
CAS nummer: 56-84-8
EINECS: 200-291-6
Merck Index: 14,840