D-(-)-Aspartic acid ≥99%
Leverantör: Thermo Scientific
Synonyms: D(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-Aspartic acid, (R)-2-Aminosuccinic acid, H-D-Asp-OH
B21184.06 B21184.14 B21184.22
B21184.06EA 675 SEK
B21184.06 B21184.14 B21184.22
D-(-)-Aspartic acid ≥99%
D-(-)-Aspartic acid
Purity: 99% CAS No.: 1783-96-6 Molecular Formula: C4H7NO4 Formula Weight: 133.10 Melting Point: 300° dec. Beilstein Reference: 1723529 Merck Reference: 14,840 Optical Rotation: -25° (c=1 in 1N HCl) MDL No.: MFCD00063081
Formel: C₄H₇NO₄
MW: 133,1 g/mol
Smältpunkt: >300 °C (dec.)
Densitet: 1,66
Storage Temperature: Omgivande
MDL Number: MFCD00063081
CAS nummer: 1783-96-6
EINECS: 217-234-6
Merck Index: 14,840