L(+)-Lactic acid, anhydrous 98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: (S)-Lactic acid, L-Lactic acid, (S)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropionic acid, (S)-(+)-Lactic acid
L13242.06 L13242.14 L13242.22
L13242.06EA 1800 SEK
L13242.06 L13242.14 L13242.22
L(+)-Lactic acid, anhydrous 98%
L(+)-Lactic acid
Purity: 98% CAS No.: 79-33-4 Molecular Formula: C3H6O3 Formula Weight: 90.08 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Odour: Odourless pH: 2.8 Melting Point: 52-54° Boiling Point: 119°/12mm Beilstein Reference: 1720251 Merck Reference: 14,5337 MDL No.: MFCD00064266
Formel: C₃H₆O₃
MW: 90.08 g/mol
Kokpunkt: 119 °C (12 mmHg)
Smältpunkt: 52…54 °C
Densitet: 1.22
Flampunkt: >110 °C (230 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00064266
CAS nummer: 79-33-4
EINECS: 201-196-2
Merck Index: 14,5337