L-Argininhydroklorid, Millipore®
Leverantör: Merck
Synonyms: L (+)-Argininhydroklorid
M-Clarity™ quality level = MQ100
1.01543.1000 1.01543.9010
1.01543.1000EA 3875 SEK
1.01543.1000 1.01543.9010
L-Argininhydroklorid, Millipore®
L-Argininhydroklorid
Formel: C₆H₁₅ClN₄O₂
MW: 210.66 g/mol
Smältpunkt: 218
Densitet: 1.42 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00064550
CAS nummer: 1119-34-2
EINECS: 214-275-1
Merck Index: 13,00785