L-Argininhydroklorid ≥98%
Leverantör: Thermo Scientific
Synonyms: L (+)-Argininhydroklorid
A14730.0E A14730.22 A14730.36
A14730.0EEA 12250 SEK
A14730.0E A14730.22 A14730.36
L-Argininhydroklorid ≥98%
L-Argininhydroklorid
Purity: 98+% CAS No.: 1119-34-2 Molecular Formula: C6H14N4O2.HCl Formula Weight: 210.67 Melting Point: 216° dec. Beilstein Reference: 3631658 Merck Reference: 14,780 Optical Rotation: +22° (c=5 in 5N HCl) MDL No.: MFCD00064550
Formel: C₆H₁₅ClN₄O₂
MW: 210,66 g/mol
Smältpunkt: 216 °C (dec.)
Densitet: 1,42
Storage Temperature: Omgivande
MDL Number: MFCD00064550
CAS nummer: 1119-34-2
EINECS: 214-275-1
Merck Index: 14,780