D-(-)-Diethyl tartrate ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: (2S,3S)-Diethyl 2,3-dihydroxysuccinate, (S,S)-(-)-Diethyl tartrate, (2S,3S)-(-)-Diethyl tartrate, (-)-Diethyl D-tartrate, (S,S)-(-)-Tartaric acid diethyl ester, D(-)-Tartaric acid diethyl ester
A17992.06 A17992.14 A17992.22
A17992.06EA 565 SEK
A17992.06 A17992.14 A17992.22
D-(-)-Diethyl tartrate ≥99%
D-(-)-Diethyl tartrate
Purity: 99% CAS No.: 13811-71-7 Molecular Formula: C8H14O6 Formula Weight: 206.19 Flash Point: 93°(199°F) Refractive Index: 1.4460 Melting Point: 17° Boiling Point: 280° Beilstein Reference: 1727143 Optical Rotation: -26.5° (c=1 in water) MDL No.: MFCD00064451
Formel: C₈H₁₄O₆
MW: 206,2 g/mol
Kokpunkt: 280 °C
Smältpunkt: 17 °C
Densitet: 1,205
Flampunkt: 93 °C (199 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00064451
CAS nummer: 13811-71-7
EINECS: 237-458-8