L (+)-asparagin, vattenfri 99%
Leverantör: Thermo Scientific
Synonyms: L-Asparagine, (S)-(+)-Asparagine, H-Asn-OH
B21473.0E B21473.22 B21473.36
B21473.0EEA 25100 SEK
B21473.0E B21473.22 B21473.36
L (+)-asparagin, vattenfri 99%
L (+)-asparagin
Purity: 99% CAS No.: 70-47-3 Molecular Formula: C4H8N2O3 Formula Weight: 132.12 Melting Point: 234-236° dec. Beilstein Reference: 1723527 Merck Reference: 14,837 MDL No.: MFCD00064401
Formel: H₂NCOCH₂CH(NH₂)CO₂H
MW: 132,12 g/mol
Kokpunkt: 343 °C (1013 hPa)
Smältpunkt: 234…236 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00064401
CAS nummer: 70-47-3
EINECS: 200-735-9
Merck Index: 14,837
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f3bc8a15c3e973916b0a6ca62d12f6953e5a027&vid=ae7eeb4ed7bc15e8e9279a950fbaf4c5111a1f46&prid=web&clid=SAPDAM