(S)-(-)-2-Klorpropionsyra 98%
Leverantör: Thermo Scientific
Fara
Synonyms: (S)-(-)-2-Chloropropanoic acid
L09427.06 L09427.14
L09427.06EA 1250 SEK
L09427.06 L09427.14
(S)-(-)-2-Klorpropionsyra 98%
(S)-(-)-2-Klorpropionsyra
Purity: 99% CAS No.: 29617-66-1 Molecular Formula: C3H5ClO2 Formula Weight: 108.53 Flash Point: 107°(225°F) Refractive Index: 1.4345 Melting Point: 4° Boiling Point: 187° Beilstein Reference: 1720257 UN No.: 2511 MDL No.: MFCD00064205
Formel: H₃CCHClCOOH
MW: 108,52 g/mol
Kokpunkt: 187 °C
Smältpunkt: 4 °C
Densitet: 1,270
Flampunkt: 107 °C (225 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00064205
CAS nummer: 29617-66-1
EINECS: 411-150-5
UN: 2511
ADR: 8,III