Järn(II)sulfat, heptahydrat ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: Iron vitriol, Ferrous sulphate heptahydrate
A15178.0C A15178.0E A15178.36
A15178.0CEA 6975 SEK
A15178.0C A15178.0E A15178.36
Järn(II)sulfat, heptahydrat ≥98%
Järn(II)sulfat, heptahydrat
Purity: 98% CAS No.: 7782-63-0 Molecular Formula: FeO4S.7H2O Formula Weight: 278.02 (151.91anhy) Melting Point: 64° -6H2O Merck Reference: 14,4057 MDL No.: MFCD00149719
Formel: FeSO₄·7H₂O
MW: 278,02 g/mol
Smältpunkt: 64 °C (-6H₂O)
Densitet: 1,897
Storage Temperature: Omgivande
MDL Number: MFCD00149719
CAS nummer: 7782-63-0
EINECS: 231-753-5
Merck Index: 14,4057