Järn (III) nitrat nonahydrat ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: Ferric nitrate nonahydrate, Iron trinitrate nonahydrate, Iron(III) nitrate hydrate, Ferric Nitrate, 9-Hydrate
12226.22 12226.A1 12226.A7
12226.22EA 615 SEK
12226.22 12226.A1 12226.A7
Järn (III) nitrat nonahydrat ≥98%
Järn (III) nitrat nonahydrat
Purity: 98+% CAS No.: 7782-61-8 Molecular Formula: FeN3O9.9H2O Formula Weight: 403.99 (241.86anhy) Physical Form: Crystalline, Contains excess HNO3 Density: 1.684 Appearance: Pale purple Odour: Acidic Melting Point: 47° Boiling Point: 125° dec. Merck Reference: 14,4027 UN No.: 1466 MDL No.: MFCD00149708
Formel: Fe(NO₃)₃·9H₂O
MW: 404 g/mol
Kokpunkt: 125 °C (dec.)
Smältpunkt: 47 °C
Densitet: 1,684
Storage Temperature: Omgivande
MDL Number: MFCD00149708
CAS nummer: 7782-61-8
EINECS: 233-899-5
UN: 1466
ADR: 5.1,III
Merck Index: 14,4027