Järn (III) nitrat nonahydrat ≥99.999%, Puratronic®
Leverantör: Thermo Fisher Scientific
Fara
10715.06 10715.14
10715.06EA 3075 SEK
10715.06 10715.14
Järn (III) nitrat nonahydrat ≥99.999%, Puratronic®
Järn (III) nitrat nonahydrat
Purity: 99.999% CAS No.: 7782-61-8 Molecular Formula: Fe(NO3)3.xH2O (x~9) Formula Weight: 241.86(anhy) Physical Form: Crystalline Aggregates Density: 1.684 Appearance: Pale purple Odour: Acidic Melting Point: 47° Boiling Point: 125° dec. Merck Reference: 14,4027 UN No.: 1466 MDL No.: MFCD00149708
Formel: Fe(NO₃)₃·9H₂O
Kokpunkt: 125 °C (dec.)
Smältpunkt: 47 °C
Densitet: 1,684
Storage Temperature: Omgivande
MDL Number: MFCD00149708
CAS nummer: 7782-61-8
EINECS: 233-899-5
UN: 1466
ADR: 5.1,III
Merck Index: 14,4027