Nickeldiklorid hexahydrat
Leverantör: MP Biomedicals
Fara
Synonyms: Nickel dichloride hexahydrate
0215582591 0215582590
ICNA0215582591EA 1950 SEK
ICNA0215582591 ICNA0215582590
Nickeldiklorid hexahydrat
Nickeldiklorid hexahydrat
Soluble in water(100 mg/ml) and alcohol.
Formel: NiCl₂·6H₂O
MW: 237,69 g/mol
Smältpunkt: 140 °C
Densitet: 1,84 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149809
CAS nummer: 7791-20-0
EINECS: 231-743-0
UN: 3288
ADR: 6.1,III
Merck Index: 13,06529