Järn(III)klorid hexahydrat 99+%, extra pure
Leverantör: Thermo Fisher Scientific
Fara
125030010 125030025 125030050 125030250
ACRO125030010EA 2775 SEK
ACRO125030010 ACRO125030025 ACRO125030050 ACRO125030250
Järn(III)klorid hexahydrat 99+%, extra pure
Järn(III)klorid hexahydrat
Formel: FeCl₃·6H₂O
Kokpunkt: 280…285 °C
Smältpunkt: 37 °C
Densitet: 1,82 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149712
CAS nummer: 10025-77-1
EINECS: 231-729-4
UN: 3260
ADR: 8,III
Merck Index: 13,04048
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=808c75376e55f29296d58eb1e5a1e184039ce733&vid=6d820873011bee3ff401441544f7f85599e40626&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Yellow to brown Granules or lumps
Titration Iodometric ≥99.0 %
Sulfate (SO4) ≤1500 ppm
Ferrous iron (Fe 2+) ≤0.9 %
Acidity (HCl) ≤0.8 %
Insoluble matter ≤0.05 % (10 g in 50 ml H2O:HCl (49:1))