Koboltdiklorid hexahydrat ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: Cobalt Chloride, Hexahydrate, Cobalt dichloride hexahydrate, Cobalt Chloride, 6-Hydrate, Cobaltous chloride hexahydrate
A16346.0B A16346.18 A16346.30
A16346.0BEA 7025 SEK
A16346.0B A16346.18 A16346.30
Koboltdiklorid hexahydrat ≥98%
Koboltdiklorid hexahydrat
Purity: 98% CAS No.: 7791-13-1 Molecular Formula: Cl2Co.6H2O Formula Weight: 237.93 (129.84anhy) Odour: Odourless pH: 4.9 Melting Point: 87° Merck Reference: 14,2437 UN No.: 3077 MDL No.: MFCD00149652
Formel: CoCl₂·6H₂O
MW: 237,93 g/mol
Kokpunkt: 110 °C (1013 hPa)
Smältpunkt: 87 °C
Densitet: 1,924
Storage Temperature: Omgivande
MDL Number: MFCD00149652
CAS nummer: 7791-13-1
EINECS: 231-589-4
UN: 3077
ADR: 9,III
Merck Index: 14,2437