Mangan(II)klorid tetrahydrat ≥99.99%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Manganese dichloride tetrahydrate, Manganese chloride (tetrahydrate), Manganese(II) chloride tetrahydrate, Manganous chloride tetrahydrate, Manganese chloride tetrahydrate, Manganese Chloride, Tetrahydrate
44442.06 44442.14 44442.22
44442.06EA 1025 SEK
44442.06 44442.14 44442.22
Mangan(II)klorid tetrahydrat ≥99.99%
Mangan(II)klorid tetrahydrat
Purity: 99.99% CAS No.: 13446-34-9 Molecular Formula: MnCl2.4H2O Formula Weight: 197.90 (125.84anhy) Physical Form: Crystalline Density: 2.010 Melting Point: 58° Merck Reference: 14,5728 MDL No.: MFCD00149792
Formel: MnCl₂·4H₂O
MW: 197.9 g/mol
Kokpunkt: 1190
Smältpunkt: 58 °C
Densitet: 2.010
Storage Temperature: Omgivande
MDL Number: MFCD00149792
CAS nummer: 13446-34-9
EINECS: 231-869-6
UN: 3077
ADR: 9 III
Merck Index: 14,5728