Mangan(II)klorid tetrahydrat ≥99.999%, Puratronic®
Leverantör: Thermo Scientific
Fara
Synonyms: Manganese dichloride tetrahydrate, Manganese chloride (tetrahydrate), Manganese(II) chloride tetrahydrate, Manganous chloride tetrahydrate, Manganese chloride tetrahydrate, Manganese Chloride, Tetrahydrate
10804.09 10804.18
10804.09EA 2425 SEK
10804.09 10804.18
Mangan(II)klorid tetrahydrat ≥99.999%, Puratronic®
Mangan(II)klorid tetrahydrat
Purity: 99.999% CAS No.: 13446-34-9 Molecular Formula: MnCl2.4H2O Formula Weight: 197.90 (125.84anhy) Physical Form: Crystalline Powder Density: 2.010 Melting Point: 58° Merck Reference: 14,5728 MDL No.: MFCD00149792
Formel: MnCl₂·4H₂O
MW: 197,9 g/mol
Kokpunkt: 1190 °C (1013 hPa)
Smältpunkt: 58 °C
Densitet: 2,010
Storage Temperature: Omgivande
MDL Number: MFCD00149792
CAS nummer: 13446-34-9
EINECS: 231-869-6
UN: 3077
ADR: 9,III
Merck Index: 14,5728