Yttrium(III)hexahydrat ≥99.9%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Yttrium trinitrate hexahydrate
12898.06 12898.14 12898.22 12898.59
12898.06EA 890 SEK
12898.06 12898.14 12898.22 12898.59
Yttrium(III)hexahydrat ≥99.9%
Yttrium(III)hexahydrat
Purity: 99.9% CAS No.: 13494-98-9 Molecular Formula: Y(NO3)3.6H2O Formula Weight: 383.01 (274.92anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Merck Reference: 14,10107 UN No.: 1477 MDL No.: MFCD00149940
Formel: N₃O₉Y·6H₂O
MW: 383.01 g/mol
Smältpunkt: 51.8
Densitet: 2.682 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149940
CAS nummer: 13494-98-9
EINECS: 233-802-6
UN: 1477
ADR: 5.1 III