Yttrium(III)hexahydrat 99.9%
Leverantör: Thermo Scientific
Varning
Synonyms: Yttrium trinitrate hexahydrate
218260250 218261000 218265000
ACRO218260250EA 1025 SEK
ACRO218260250 ACRO218261000 ACRO218265000
Yttrium(III)hexahydrat 99.9%
Yttrium(III)hexahydrat
Formel: N₃O₉Y·6H₂O
MW: 383,01 g/mol
Smältpunkt: 51,8 °C
Densitet: 2,682 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149940
CAS nummer: 13494-98-9
EINECS: 233-802-6
UN: 3077
ADR: 9,III
Order Now
Specifikation testresultat
Appearance White Adhering crystalline powder or crystals
Total trace metal impurities ≤0.1 %