Cadmium(II) sulphate hydrate ≥99.996%, Puratronic®
Leverantör: Thermo Fisher Scientific
Fara
10675.09 10675.18
10675.09EA 1750 SEK
10675.09 10675.18
Cadmium(II) sulphate hydrate ≥99.996%, Puratronic®
Cadmium(II) sulphate hydrate
Purity: 99.996% CAS No.: 15244-35-6 Molecular Formula: 3CdSO4.xH2O (x~8) Formula Weight: 208.46(anhy) Physical Form: Crystalline Density: 3.08 Refractive Index: 1.565 Appearance: White Odour: Odourless Melting Point: 40° -H2O Merck Reference: 14,1627 UN No.: 2570 MDL No.: MFCD00151655
Formel: CdO₄S·H₂O
MW: 226.49 g/mol
Smältpunkt: >40 °C (-H₂O)
Densitet: 3.08
Storage Temperature: Omgivande
MDL Number: MFCD00151655
CAS nummer: 15244-35-6
EINECS: 233-331-6
UN: 2570
ADR: 6.1 III
Merck Index: 14,1627