Kadmium (II) klorid hemipentahydrate 79.5-81.0% ACS
Leverantör: Thermo Fisher Scientific
Fara
12373.18 12373.30
12373.18EA 1325 SEK
12373.18 12373.30
Kadmium (II) klorid hemipentahydrate 79.5-81.0% ACS
Kadmium (II) klorid hemipentahydrate
CAS No.: 7790-78-5 Molecular Formula: CdCl2.2.5H2O Formula Weight: 228.36 (183.32anhy) Physical Form: Crystalline Refractive Index: 1.6513 Sensitivity: Hygroscopic Merck Reference: 14,1617 UN No.: 2570 MDL No.: MFCD00799071
Formel: CdCl₂·2.5H₂O
MW: 228.35 g/mol
Smältpunkt: 568
Densitet: 2.84 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00799071
CAS nummer: 7790-78-5
EINECS: 233-296-7
UN: 2570
ADR: 6.1 III
Merck Index: 14,1617