Koppar(II)sulfat pentahydrat, Supelco®
Leverantör: Merck
Fara
M-Clarity™ quality level = MQ200
1.02780.5000 1.02780.9050
1.02780.5000EA 6270 SEK
1.02780.5000 1.02780.9050
Koppar(II)sulfat pentahydrat, Supelco®
Koppar(II)sulfat pentahydrat
Formel: CuSO₄·5H₂O
Densitet: 2,284 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149681
CAS nummer: 7758-99-8
EINECS: 231-847-6
UN: 3077
ADR: 9,III
REACH: 01-2119520566-40
Merck Index: 13,02682