Koppar(II)sulfat pentahydrat ≥99%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Cupric sulphate pentahydrate, Copper sulphate pentahydrate
A11262.0B A11262.0G A11262.0I
A11262.0BEA 1050 SEK
A11262.0B A11262.0G A11262.0I
Koppar(II)sulfat pentahydrat ≥99%
Koppar(II)sulfat pentahydrat
Purity: 99% CAS No.: 7758-99-8 Molecular Formula: CuSO4.5H2O Formula Weight: 249.68 (159.60anhy) Odour: Odourless pH: 4 Merck Reference: 14,2653 UN No.: 3288 MDL No.: MFCD00149681
Formel: CuSO₄·5H₂O
MW: 249.69 g/mol
Densitet: 2.270
Storage Temperature: Omgivande
MDL Number: MFCD00149681
CAS nummer: 7758-99-8
EINECS: 231-847-6
UN: 3288
ADR: 6.1 III
REACH: 01-2119520566-40
Merck Index: 13,02682