Samariumtrinitrat hexahydrat 99,99 REO basis, REacton®
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Samarium (III) nitrate hexahydrate, Samarium(III) nitrate hydrate
11224.06 11224.14 11224.22
11224.06EA 1175 SEK
11224.06 11224.14 11224.22
Samariumtrinitrat hexahydrat 99,99 REO basis, REacton®
Samariumtrinitrat hexahydrat
Purity: 99.99% CAS No.: 13759-83-6 Molecular Formula: Sm(NO3)3.xH2O (x≈6) Formula Weight: 336.36(anhy) Physical Form: Crystalline Aggregates Density: 2.375 Appearance: Pale yellow Odour: Odourless pH: ca. 3 Melting Point: 78-79° UN No.: 1477 MDL No.: MFCD00149857
Formel: N₃O₉Sm·6H₂O
MW: 444.47 g/mol
Smältpunkt: 78…79 °C
Densitet: 2.375
Storage Temperature: Omgivande
MDL Number: MFCD00149857
CAS nummer: 13759-83-6
EINECS: 233-798-6
UN: 1477
ADR: 5.1 III