Erbium trinitrate pentahydrate ≥99.9% (REO, rare earth oxide basis)
Leverantör: Thermo Scientific
Fara
Synonyms: Erbium(III) nitrate hydrate, Erbium (III) nitrate pentahydrate
12920.14 12920.22
12920.14EA 2775 SEK
12920.14 12920.22
Erbium trinitrate pentahydrate ≥99.9% (REO, rare earth oxide basis)
Erbium trinitrate pentahydrate
Purity: 99.9% Molecular Formula: Er(NO3)3.xH2O (x≈5) Formula Weight: 353.27(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Melting Point: 130° -4H2O UN No.: 1477 MDL No.: MFCD00010988
Formel: ErN₃O₉·5H₂O
MW: 443,35 g/mol
Smältpunkt: 130 °C (-4H₂O)
Storage Temperature: Omgivande
MDL Number: MFCD00149691
CAS nummer: 10031-51-3
EINECS: 233-436-7
UN: 1477
ADR: 5.1,III