Gadolinium(III)hexahydrat ≥99.99% (REO, rare earth oxide basis), REacton®
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Gadolinium trichloride hexahydrate
11287.06 11287.14 11287.22
11287.06EA 2275 SEK
11287.06 11287.14 11287.22
Gadolinium(III)hexahydrat ≥99.99% (REO, rare earth oxide basis), REacton®
Gadolinium(III)hexahydrat
Purity: 99.99% CAS No.: 13450-84-5 Molecular Formula: GdCl3.6H2O Formula Weight: 371.70 (263.61anhy) Physical Form: Crystalline Aggregates Density: 2.424 Appearance: White Odour: Characteristic Merck Reference: 14,4326 MDL No.: MFCD00149732
Formel: Cl₃Gd·6H₂O
MW: 371.7 g/mol
Densitet: 2.424
Storage Temperature: Omgivande
MDL Number: MFCD00149732
CAS nummer: 13450-84-5
EINECS: 233-386-6
Merck Index: 14,4326