Terbium(III) bromide hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Leverantör: Thermo Fisher Scientific
13173.09 13173.18
13173.09EA 3650 SEK
13173.09 13173.18
Terbium(III) bromide hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Terbium(III) bromide hydrate
Purity: 99.99% Molecular Formula: TbBr3.xH2O Formula Weight: 398.65(anhy) Physical Form: Crystalline Melting Point: 170° dec. MDL No.: MFCD00049571
Formel: Br₃Tb·H₂O
MW: 416.65 g/mol
Smältpunkt: 170 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00049571
CAS nummer: 15162-98-8
EINECS: 238-442-3