Samarium tribromide hexahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Leverantör: Thermo Scientific
Varning
Synonyms: Samarium tribromide hexahydrate, Samarium(III) bromide hexahydrate
14603.14 14603.22
14603.14EA 2200 SEK
14603.14 14603.22
Samarium tribromide hexahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Samarium tribromide hexahydrate
Purity: 99.9% CAS No.: 13517-12-9 Molecular Formula: SmBr3.6H2O Formula Weight: 498.15 (390.06anhy) Physical Form: Crystalline Sensitivity: Hygroscopic MDL No.: MFCD00192430
Formel: Br₃Sm·6H₂O
MW: 498,16 g/mol
Smältpunkt: >640 °C
Storage Temperature: Omgivande
MDL Number: MFCD00192430
CAS nummer: 13517-12-9
EINECS: 237-347-4