Ammoniumjärn(II)sulfat hexahydrat ≥99.999%, Puratronic®
Leverantör: Thermo Fisher Scientific
Varning
40751.03 40751.06 40751.14
40751.03EA 920 SEK
40751.03 40751.06 40751.14
Ammoniumjärn(II)sulfat hexahydrat ≥99.999%, Puratronic®
Ammoniumjärn(II)sulfat hexahydrat
Purity: 99.999% CAS No.: 7783-85-9 Molecular Formula: H8FeN2O8S2.xH2O (x≈6) Formula Weight: 284.04(anhy) Physical Form: Coarse Powder Density: 1.864 Appearance: Pale bluish green Odour: Odourless Melting Point: ca 100° dec. Merck Reference: 14,521 MDL No.: MFCD00150530
Formel: (NH₄)₂Fe(SO₄)₂·6H₂O
Smältpunkt: ∼ 100 °C (dec.)
Densitet: 1,864
Storage Temperature: Omgivande
MDL Number: MFCD00150530
CAS nummer: 7783-85-9
EINECS: 233-382-4
Merck Index: 14,521