Tetramethylammonium hydroxide pentahydrate ≥98%
Leverantör: Thermo Fisher Scientific
Fara
L09658.14 L09658.22
L09658.14EA 1250 SEK
L09658.14 L09658.22
Tetramethylammonium hydroxide pentahydrate ≥98%
Tetramethylammonium hydroxide pentahydrate
Purity: 98% CAS No.: 10424-65-4 Molecular Formula: C4H13NO.5H2O Formula Weight: 181.23 (91.15anhy) Melting Point: 66-67° Beilstein Reference: 3714235 Merck Reference: 14,9224 UN No.: 3423 MDL No.: MFCD00149566
Formel: C₄H₁₃NO·5H₂O
Smältpunkt: 66…67 °C
Densitet: 1,829 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149566
CAS nummer: 10424-65-4
EINECS: 200-882-9
UN: 3423
ADR: 8,II
Merck Index: 14,9224