Magnesiumsulfat heptahydrat ≥99%
Leverantör: Thermo Fisher Scientific
Synonyms: sulphuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt
A14491.0B A14491.0I
A14491.0BEA 770 SEK
A14491.0B A14491.0I
Magnesiumsulfat heptahydrat ≥99%
Magnesiumsulfat heptahydrat
Purity: 99+% CAS No.: 10034-99-8 Molecular Formula: MgSO4.7H2O Formula Weight: 246.48 (120.37anhy) Odour: Odourless Merck Reference: 14,5691 MDL No.: MFCD00149785
Formel: MgSO₄·7H₂O
MW: 246.48 g/mol
Smältpunkt: 1124
Densitet: 1.68
Storage Temperature: Omgivande
MDL Number: MFCD00149785
CAS nummer: 10034-99-8
EINECS: 231-298-2
Merck Index: 13,05715