Kalciumsulfat hemihydrat 97+%, ren
Leverantör: Thermo Fisher Scientific
Synonyms: Plaster of Paris
385351000 385355000 385350025
ACRO385351000EA 440 SEK
ACRO385351000 ACRO385355000 ACRO385350025
Kalciumsulfat hemihydrat 97+%, ren
Kalciumsulfat hemihydrat
Formel: CaSO₄·0.5H₂O
Smältpunkt: ? 1400 °C
Densitet: 2,73 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149624
CAS nummer: 10034-76-1
EINECS: 231-900-3
Order Now
Specifikation testresultat
Appearance White to off-white Powder
Titration Complexometric ≥97.0 % (On dry substance)
Identification Confirmed
Loss on ignition 4.5 % to 8.0 % (500°C, 2 h)