Berylliumsulfat tetrahydrat ≥99.99%
Leverantör: Thermo Scientific
Fara
16104.14 16104.22
16104.14EA 1775 SEK
16104.14 16104.22
Berylliumsulfat tetrahydrat ≥99.99%
Berylliumsulfat tetrahydrat
Purity: 99.99% CAS No.: 7787-56-6 Molecular Formula: BeSO4.4H2O Formula Weight: 177.14 (105.07anhy) Physical Form: Lump Density: 1.1713 Solubility: Freely soluble in water. Insoluble in alcohol Merck Reference: 14,1175 UN No.: 1566 MDL No.: MFCD00149156
Formel: BeO₄S·4H₂O
MW: 177,14 g/mol
Kokpunkt: 580 °C (1013 hPa)
Smältpunkt: 110 °C
Densitet: 1,1713
Storage Temperature: Omgivande
MDL Number: MFCD00149156
CAS nummer: 7787-56-6
EINECS: 236-842-2
UN: 1566
ADR: 6.1,II
Merck Index: 14,1175