Capping A (Tetrahydrofurane:2,6-Lutidine:Acetic anhydride) (8:1:1) v/v/v for DNA/RNA synthesis
Leverantör: Merck
Fara
M-Clarity™ quality level = MQ200
1.18603.0450 1.18603.2500
1.18603.0450EA 961 SEK
1.18603.0450 1.18603.2500
Capping A (Tetrahydrofurane:2,6-Lutidine:Acetic anhydride) (8:1:1) v/v/v for DNA/RNA synthesis
Capping Reagent
Flampunkt: 4 °C
UN: 2924
ADR: 3 (8),