Magnesiumnitrat hexahydrat ≥99.97%
Leverantör: Thermo Fisher Scientific
Varning
87609.06 87609.18 87609.30
87609.06EA 785 SEK
87609.06 87609.18 87609.30
Magnesiumnitrat hexahydrat ≥99.97%
Magnesiumnitrat hexahydrat
Purity: 99.97% CAS No.: 13446-18-9 Molecular Formula: Mg(NO3)2.6H2O Formula Weight: 256.41 (148.32anhy) Physical Form: Crystalline Density: 1.46 Melting Point: 89° Boiling Point: 330° dec. Merck Reference: 14,5674 UN No.: 1474 MDL No.: MFCD00149779
Formel: Mg(NO₃)₂·6H₂O
Kokpunkt: 330 °C (dec.)
Smältpunkt: 89 °C
Densitet: 1,46
Storage Temperature: Omgivande
MDL Number: MFCD00149779
CAS nummer: 13446-18-9
EINECS: 233-826-7
UN: 1474
ADR: 5.1,III
Merck Index: 14,5674