Magnesiumnitrat hexahydrat 98.0-102.0% ACS
Leverantör: Thermo Fisher Scientific
Varning
11564.18 11564.36 11564.A3
11564.18EA 725 SEK
11564.18 11564.36 11564.A3
Magnesiumnitrat hexahydrat 98.0-102.0% ACS
Magnesiumnitrat hexahydrat
Purity: 98.0-102.0% CAS No.: 13446-18-9 Molecular Formula: Mg(NO3)2.6H2O Formula Weight: 256.41 (148.32anhy) Physical Form: Crystalline Density: 1.46 Melting Point: 89° Boiling Point: 330° dec. Solubility: Soluble in water, alcohol and liquid NH3 Merck Reference: 14,5674 UN No.: 1474 MDL No.: MFCD00149779
As fertilizer, in manufacture of ammonium nitrate, as a reagent chemical, and in pyrotechnics
Formel: Mg(NO₃)₂·6H₂O
MW: 256.41 g/mol
Kokpunkt: 330 °C (dec.)
Smältpunkt: 89
Densitet: 1.46
Flampunkt: 93,3
Storage Temperature: Omgivande
MDL Number: MFCD00149779
CAS nummer: 13446-18-9
EINECS: 233-826-7
UN: 1474
ADR: 5.1 III
Merck Index: 14,5674