Krom(III)kaliumsulfat dodekahydratet ≥99.9%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Chromium potassium sulphate dodecahydrate, Chromium (III) potassium bis(sulphate) dodecahydrate
44083.14 44083.22
44083.14EA 1925 SEK
44083.14 44083.22
Krom(III)kaliumsulfat dodekahydratet ≥99.9%
Krom(III)kaliumsulfat dodekahydratet
Purity: 99.9% CAS No.: 7788-99-0 Molecular Formula: CrKO8S2.12H2O Formula Weight: 499.39 (283.21anhy) Physical Form: Crystalline Density: 1.83 Appearance: Dark purple Odour: Odourless Melting Point: 89° Merck Reference: 14,2229 MDL No.: MFCD00149917
Formel: CrK(SO₄)₂·12H₂O
MW: 499.4 g/mol
Smältpunkt: 89
Densitet: 1.83
Storage Temperature: Omgivande
MDL Number: MFCD00149917
CAS nummer: 7788-99-0
EINECS: 233-401-6
Merck Index: 14,2229