di-Natriumhydrogen fosfat heptahydrat 98.0-102.0% ACS
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Sodium phosphate dibasic heptahydrate, Sodium hydrogen phosphate heptahydrate, Sodium monohydrogen phosphate heptahydrate
11592.36 11592.A3
11592.36EA 1725 SEK
11592.36 11592.A3
di-Natriumhydrogen fosfat heptahydrat 98.0-102.0% ACS
di-Natriumhydrogen fosfat heptahydrat
Purity: 98.0-102.0% CAS No.: 7782-85-6 Molecular Formula: Na2HPO4.7H2O Formula Weight: 268.07 (141.98anhy) Physical Form: Crystalline Density: 1.7 Appearance: White Odour: Odourless Melting Point: 48.1°-5H2O Merck Reference: 14,8659 MDL No.: MFCD00149180
Formel: HNa₂O₄P·7H₂O
MW: 268.07 g/mol
Smältpunkt: 48.1 °C (-5H₂O)
Densitet: 1.7
Storage Temperature: Omgivande
MDL Number: MFCD00149180
CAS nummer: 7782-85-6
EINECS: 231-448-7
Merck Index: 14,8659