trinatriumortofosfat dodekahydrat, BAKER, Laboratory Reagent, J.T.Baker®
Leverantör: Avantor
Varning
Synonyms: Sodium Phosphate tribasic 12-Hydrate, Sodium phosphate dodecahydrate, Sodium phosphate tribasic dodecahydrate
0308.1000EA 355 SEK
0308.1000 0308.9025BB
trinatriumortofosfat dodekahydrat, BAKER, Laboratory Reagent, J.T.Baker®
trinatriumortofosfat dodekahydrat
Formel: Na₃PO₄·12H₂O
MW: 380.12 g/mol
Smältpunkt: ∼75 °C
Densitet: 1.62 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149198
CAS nummer: 10101-89-0
EINECS: 231-509-8
REACH: 01-2119489800-32-xxxx
Merck Index: 13,08736
Order Now
Specifikation testresultat
Assay min. 98 %