Natriummolybdat(VI)dihydrat ≥98%
Leverantör: Thermo Scientific
Synonyms: Molybdic acid, sodium salt dihydrate, Molybdic acid sodium salt dihydrate, Molybdic acid disodium salt dihydrate, di-Sodium molybdate dihydrate, Sodium molybdate dihydrate, Sodium molybdenum oxide dihydrate
A19222.0E A19222.22 A19222.36
A19222.0EEA 12400 SEK
A19222.0E A19222.22 A19222.36
Natriummolybdat(VI)dihydrat ≥98%
Natriummolybdat(VI)dihydrat
Purity: 98% CAS No.: 10102-40-6 Molecular Formula: Na2MoO4.2H2O Formula Weight: 241.95 (205.92anhy) Odour: Odourless Melting Point: 100° -2H2O MDL No.: MFCD00149170
Formel: Na₂MoO₄·2H₂O
MW: 241,95 g/mol
Smältpunkt: 100 °C (-2H₂O)
Densitet: 2,71
Storage Temperature: Omgivande
MDL Number: MFCD00149170
CAS nummer: 10102-40-6
EINECS: 231-551-7
Merck Index: 14,8645