Kaliumhexacyanoferrat(II)trihydrat 98.5-102.0% ACS
Leverantör: Thermo Fisher Scientific
Synonyms: Potassiumhexacyanoferrate(II) trihydrate, Tetrapotassium hexacyanoferrate trihydrate, Potassium ferrocyanide trihydrate
33358.22 33358.36 33358.A3
33358.22EA 920 SEK
33358.22 33358.36 33358.A3
Kaliumhexacyanoferrat(II)trihydrat 98.5-102.0% ACS
Kaliumhexacyanoferrat(II)trihydrat
Purity: 98.5-102.0% CAS No.: 14459-95-1 Molecular Formula: K4Fe(CN)6.3H2O Formula Weight: 422.39 (368.36anhy) Physical Form: Crystalline Density: 1.85 Appearance: Pale yellow Odour: Odourless pH: 9.5 Melting Point: 69-71° Merck Reference: 14,7631 MDL No.: MFCD00167023
Formel: K₄Fe(CN)₆·3H₂O
MW: 422.39 g/mol
Smältpunkt: 69…71 °C
Densitet: 1.85
Storage Temperature: Omgivande
MDL Number: MFCD00167023
CAS nummer: 14459-95-1
EINECS: 237-722-2
Merck Index: 14,7631
Order Now
SPECIFIKATIONER
Environmentally Preferable