Piperazin hexahydrat 98%
Leverantör: Thermo Scientific
Fara
Synonyms: Piperazine hydrate
131300010 131301000 131305000 131300051
ACRO131300010EA 2200 SEK
ACRO131300010 ACRO131301000 ACRO131305000 ACRO131300051
Piperazin hexahydrat 98%
Piperazin hexahydrat
Formel: C₄H₁₀N₂·6H₂O
MW: 194,23 g/mol
Kokpunkt: 145…156 °C (760 mmHg)
Smältpunkt: 41…45 °C
Flampunkt: 87 °C
Storage Temperature: Omgivande
MDL Number: MFCD00149389
CAS nummer: 142-63-2
EINECS: 203-808-3
UN: 2579
ADR: 8,III
Merck Index: 12,07617
Order Now
Specifikation testresultat
Appearance Transparent to white Adhering crystals
Infrared spectrum Conforms
Titration with HCl ≥97.5 %
HPLC ≥96.0 %