3,5-Dihydroxytoluen monohydrat 99%
Leverantör: Thermo Fisher Scientific
Varning
129550250 129551000 129555000
ACRO129550250EA 1825 SEK
ACRO129550250 ACRO129551000 ACRO129555000
3,5-Dihydroxytoluen monohydrat 99%
3,5-Dihydroxytoluen monohydrat
Formel: C₇H₈O₂·1H₂O
Kokpunkt: 290 °C (760 mmHg)
Smältpunkt: 56…61 °C
Densitet: 1,29 g/cm³ (4 °C)
Flampunkt: 159 °C
Storage Temperature: Omgivande
MDL Number: MFCD00149092
CAS nummer: 6153-39-5
EINECS: 207-984-2
Merck Index: 12,06995
Order Now
Specifikation testresultat
Appearance White to beige to pink-brown Powder or crystals
Infrared spectrum Conforms
Melting point 56 to 61 °C
Water 10 to 14 % (K.F.)
Solubility (5 % in water) Clear colorless to light brown
HPLC ≥98.5 %