Natriumoktan-1-sulfonat, hydrate 99+% för HPLC
Leverantör: Thermo Fisher Scientific
Synonyms: Sodium octane-1-sulphonate
42636.06 42636.14 42636.22
42636.06EA 1675 SEK
42636.06 42636.14 42636.22
Natriumoktan-1-sulfonat, hydrate 99+% för HPLC
Natriumoktan-1-sulfonat
Ion-pair reagent for HPLC analysis
Purity: 99+% CAS No.: 5324-84-5 Molecular Formula: CH3(CH2)7SO3Na.xH2O Formula Weight: 216.28(anhy) Physical Form: Crystalline Melting Point: 300° MDL No.: MFCD00007544
Sodium-1-octanesulfonate is used as a reagent for HPLC analysis of organic compounds, and more specifically, peptides and proteins. It is also extensively used as an ion pairing agent.
Formel: C₈H₁₇NaO₃S
MW: 216.28 g/mol
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007544
CAS nummer: 5324-84-5
EINECS: 226-195-4
Order Now
Specifikation testresultat
Loss on drying 6.9-8.5%
10% water solutionClear Pass Test.
colorless 0.4% EtOH/MeOHClear Pass Test
clear0.4% EtOH/MeOH Clear
IR identification Pass test