1-Hexanesulphonic acid sodium salt monohydrate, Supelco®
Leverantör: Merck
Synonyms: Sodium 1-hexanesulphonate monohydrate
M-Clarity™ quality level = MQ100
52862-10G-F 52862-50G-F
52862-10G-FEA 1700 SEK
52862-10G-F 52862-50G-F
1-Hexanesulphonic acid sodium salt monohydrate, Supelco®
1-Hexanesulphonic acid sodium salt monohydrate
Formel: C₆H₁₃NaO₃S·1H₂O
MW: 206.24 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00149549
CAS nummer: 207300-91-2
EINECS: 220-601-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=897e40e9856d4ea56d810990e51bee4045dd30d6&vid=d9488496ad880fedc4f73526c19d1430ba156df1&prid=web&clid=SAPDAM