trinatriumcitratdihydrat, Millipore®
Leverantör: Merck Millipore Calbiochem
Synonyms: Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Sodium Citrate, Dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate
567446-1 567446-100 567446-5
567446-1EA 1675 SEK
567446-1 567446-100 567446-5
trinatriumcitratdihydrat, Millipore®
trinatriumcitratdihydrat
Formel: Na₃C₆H₅O₇·2H₂O
MW: 294.1 g/mol
Smältpunkt: 150
Densitet: 1.76 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00150031
CAS nummer: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM