Kalciumoxalat monohydrat ≥99.9985% (excluding 25 ppm max alkali earths), Puratronic®
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Oxalic acid calcium salt monohydrate, Calcium ethanedioate monohydrate
13007.06 13007.14
13007.06EA 1550 SEK
13007.06 13007.14
Kalciumoxalat monohydrat ≥99.9985% (excluding 25 ppm max alkali earths), Puratronic®
Kalciumoxalat monohydrat
Purity: 99.9985% CAS No.: 5794-28-5 Molecular Formula: CaC2O4.H2O Formula Weight: 146.11 (128.10anhy) Physical Form: Powder Density: 2.12 Melting Point: 200° Merck Reference: 14,1685 UN No.: 3288 MDL No.: MFCD00150039
Formel: C₂CaO₄·1H₂O
MW: 146.11 g/mol
Smältpunkt: 200
Densitet: 2.12
Storage Temperature: Omgivande
MDL Number: MFCD00150039
CAS nummer: 5794-28-5
EINECS: 209-260-1
UN: 3288
ADR: 6.1 III
Merck Index: 14,1685