Oxalsyra dihydrat ≥98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Ethanedioic acid dihydrate
A13866.0B A13866.0I A13866.30
A13866.0BEA 1500 SEK
A13866.0B A13866.0I A13866.30
Oxalsyra dihydrat ≥98%
Oxalsyra dihydrat
Purity: 98% CAS No.: 6153-56-6 Molecular Formula: C2H2O4.2H2O Formula Weight: 126.07 (90.04anhy) Odour: Odourless pH: 1.3 Melting Point: 101-105° Beilstein Reference: 3679436 Merck Reference: 14,6911 UN No.: 3261 MDL No.: MFCD00149102
Formel: HO₂CCO₂H·2H₂O
Kokpunkt: 149…160 °C (1013 hPa)
Smältpunkt: 101…105 °C
Densitet: 1,65
Storage Temperature: Omgivande
MDL Number: MFCD00149102
CAS nummer: 6153-56-6
EINECS: 205-634-3
UN: 3261
ADR: 8,III
REACH: 01-2119534576-33
Merck Index: 14,6911
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcc1447f5f23a589dc85e10dd090c86e33b6541b&vid=2f0e34543b2f625715b002905fc0dd45fcd75561&prid=web&clid=SAPDAM