Magnesiumacetat tetrahydrat ≥98%
Leverantör: Thermo Scientific
Synonyms: Acetic acid magnesium salt tetrahydrate, Magnesium di(acetate) tetrahydrate
12225.30 12225.A1
12225.30EA 1125 SEK
12225.30 12225.A1
Magnesiumacetat tetrahydrat ≥98%
Magnesiumacetat tetrahydrat
Formel: (H₃CCOO)₂Mg·4H₂O
MW: 214,46 g/mol
Smältpunkt: 80 °C
Densitet: 1,454
Storage Temperature: Omgivande
MDL Number: MFCD00149214
CAS nummer: 16674-78-5
EINECS: 205-554-9
Merck Index: 14,5654